Search Thermo Fisher Scientific
PA1-28389 detects IL1 receptor I from human samples.
PA1-28389 has been successfully used in immunoprecipitation, radioimmune assay applications.
The PA1-28389 immunogen is: Recombinant full length protein (Human).
Endotoxin Content: <10 pg/ml by LAL method.
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.