Invitrogen
FIGURE: 1 / 4
MA5-13764 targets Insulin Receptor alpha in ELISA, FACS, ICC/IF and IHC (P) applications and shows reactivity with Bovine, Human, Ovine, Porcine, Rat and Rabbit samples.
The MA5-13764 immunogen is iM-9 lymphocytes followed by IR.
Biological actions of insulin and IGF-1 are mediated by their respective cell surface receptor tyrosine kinases that regulate multiple signaling pathways through activation of a series of phosphorylation cascades. The insulin receptor. Insulin/IGF-1 binding to the extracellular domain leads to autophosphorylation of downstream target proteins. These two receptors differ in sequence in regions that confer specificity for the designated ligand as well as in certain intracellular signaling domains, resulting in significant differences in the functional consequences of activation of each receptor. Defects in IR are the cause of various insulin resistance syndromes and IGF-1R defects may cause some forms of growth retardation. Both these signaling cascades are also important for the development of cancer.