Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MCGSALAFFT AAFVCLQNDR RGPASFLWAA WVFSLVLGLG QGEDNRCASS NAASCARCLA LGPECGWCVQ EDFISGGSRS ERCDIVSNLI SKGCSVDSIE YPSVHVIIPT ENEINTQVTP GEVSIQLRPG AEANFMLKVH PLKKYPVDLY YLVDVSASMH NNIEKLNSVG NDLSRKMAFF SRDFRLGFGS YVDKTVSPYI SIHPERIHNQ CSDYNLDCMP PHGYIHVLSL TENITEFEKA VHRQKISGNI DTPEGGFDAM LQAAVCESHI GWRKEAKRLL LVMTDQTSHL ALDSKLAGIV VPNDGNCHLK NNVYVKSTTM EHPSLGQLSE KLIDNNINVI FAVQGKQFHW YKDLLPLLPG TIAGEIESKA ANLNNLVVEA YQKLISEVKV QVENQVQGIY FNITAICPDG SRKPGMEGCR NVTSNDEVLF NVTVTMKKCD VTGGKNYAII KPIGFNETAK IHIHRNCSCQ CEDNRGPKGK CVDETFLDSK CFQCDENKCH FDEDQFSSES CKSHKDQPVC SGRGVCVCGK CSCHKIKLGK VYGKYCEKDD FSCPYHHGNL CAGHGECEAG RCQCFSGWEG DRCQCPSAAA QHCVNSKGQV CSGRGTCVCG RCECTDPRSI GRFCEHCPTC YTACKENWNC MQCLHPHNLS QAILDQCKTS CALMEQQHYV DQTSECFSSP SYLRIFFIIF IVTFLIGLLK VLIIRQVILQ WNSNKIKSSS DYRVSASKKD KLILQSVCTR AVTYRREKPE EIKMDISKLN AHETFRCNF
ITGB8 (Integrin beta 8) is a member of the integrin beta chain family and is a single-pass type I membrane protein with a VWFA domain and four cysteine-rich repeats. This protein noncovalently binds to an alpha subunit to form a heterodimeric integrin complex. In general, integrin complexes mediate cell-cell and cell-extracellular matrix interactions and this complex plays a role in human airway epithelial proliferation. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: integrin beta 8; Integrin beta-8; Integrin beta8; integrin, beta 8
Gene Aliases: ITGB8
UniProt ID: (Human) P26012
Entrez Gene ID: (Human) 3696
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support