Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
This antibody detects endogenous protein at a molecular weight of 71 and 101 kDa.
Purity is >95% by SDS-PAGE.
The inter-alpha trypsin inhibitor (ITI) family is a group of structurally related plasma serine protease inhibitors synthesized in the liver and built up from different combinations of three highly homologous heavy chains (ITI-H1, ITI-H2 and ITI-H3) and one light chain (Bikunin). A fourth member of the ITI family, ITI-H4 (also known as I a IH4P) harbors a Pro-rich region (PRR) in its C-terminus. ITI is a glycoprotein composed of three polypeptides linked by chondroitin sulphate: two heavy chains, ITI-H1 and ITI-H2, and Bikunin. Bikunin confers the protease-inhibitor function of ITI. The heavy chains of the ITI family, designated as SHAPs (for serum-derived hyaluronan-associated proteins), bind covalently to hyaluronic acid (HA), resulting in pericellular matrix stabilization. ITI-H1 contains a potential peptide which could stimulate a broad spectrum of phagocytotic cells. Although ITI-H1, ITI-H3 and Bikunin have antitumoral and antimetastatic properties in the cell, they are also associated with malignant transformation of lung tissue. ITI-H1 and ITI-H2 are associated with calcium oxalate stone formation in kidney and urine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inter-alpha (globulin) inhibitor, H1 polypeptide; inter-alpha-inhibitor heavy chain 1; Inter-alpha-trypsin inhibitor complex component III; Inter-alpha-trypsin inhibitor heavy chain H1; ITI heavy chain H1; Serum-derived hyaluronan-associated protein; SHAP
Gene Aliases: H1P; IATIH; IGHEP1; Intin1; ITI-HC1; ITIH; Itih-1; ITIH1; SHAP
UniProt ID: (Human) P19827, (Mouse) Q61702
Entrez Gene ID: (Human) 3697, (Mouse) 16424, (Rat) 306251
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support