Search Thermo Fisher Scientific
PA1-27201 detects Influenza A Virus from virus samples. Specific to H1N1 by IHA. Does not react with Hep-2 cells. May cross-react with chicken cellular proteins. Does not cross-react with Influenza B, RSV, Para1-3 or Adenovirus.
PA1-27201 has been successfully used in immunohistochemistry (paraffin) and immunofluorescence procedures. For immunohistochemistry, acetone fixation of the antigen source is recommended prior to staining.
The PA1-27201 immunogen is Influenza A strain USSR (H1N1)
Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.