Search Thermo Fisher Scientific
Heat mediated antigen retrieval recommended prior to tissue staining.
This antibody does not react with mouse.
This antibody is predicted to react with rat based on sequence homology.
Component of the AP-1 transcription factor that represses transactivation mediated by the Jun family of proteins. Involved in a variety of transcriptional responses associated with AP-1 such as UV-induced apoptosis, cell differentiation, tumorigenesis and antitumogeneris. Can also function as a repressor by recruiting histone deacetylase 3/HDAC3 to the promoter region of JUN. May control transcription via direct regulation of the modification of histones and the assembly of chromatin.