Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:3,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:100-1:1,000 | - |
Immunocytochemistry (ICC/IF) |
1:100-1:1,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Recombinant fragment corresponding to a region within amino acids 5 and 278 of Human JNK3 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2545326 |
PA5-27850 targets JNK3 in IHC (P) and WB applications and shows reactivity with Human samples.
The PA5-27850 immunogen is recombinant fragment corresponding to a region within amino acids 5 and 278 of Human JNK3.
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This protein is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). Through its phosphorylation and nuclear localization, this kinase plays regulatory roles in the signaling pathways during neuronal apoptosis. Beta-arrestin 2, a receptor-regulated MAP kinase scaffold protein, is found to interact with, and stimulate the phosphorylation of this kinase by MAP kinase kinase 4 (NhpK4). Cyclin-dependent kianse 5 can phosphorylate, and inhibit the activity of this kinase, which may be important in preventing neuronal apoptosis. Four alternatively spliced transcript variants encoding distinct isoforms have been reported.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Jun N-terminal kinase 3; JNK3 alpha protein kinase; MAP kinase 10; MAP kinase p49 3F12; MAPK 10; Mitogen-activated protein kinase 10; OTTHUMP00000161181; OTTHUMP00000161183; OTTHUMP00000218718; SAPK1b; stress activated protein kinase beta; Stress-activated protein kinase 1b; Stress-activated protein kinase JNK3
Gene Aliases: JNK3; JNK3A; MAPK10; PRKM10; SAPK1B
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support