Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 175 and 225 |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
0.10 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
TBS with 0.1% BSA |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat
The KRAB (Kruppel-associated box) domain is about 45 amino acids in length and is a transcriptional repression domain found in numerous transcription factors. There have been identified over 220 KRAB-zinc finger protein (KRAB-ZFP) genes in the human genome. These proteins functionally repress transcription via specific interactions with KAP-1 (KRAB-associated protein 1). KAP-1 is an 835 amino acid polypeptide that contains a RING finger, B boxes, and a PHD finger. KAP-1 has been shown to form complexes with KRAB-domain transcription factors and increase the efficiency with which they mediate repression. KAP-1 has also been shown to directly interact with HP1 (heterochromatin protein 1) and KRAZ1 (Kruppel-associated box-containing zinc finger protein 1). KAP-1 directly targets KRAZ1 to the foci of centromeric heterochromatin containing HP1alpha, thus helping to regulate transcriptional repression. Studies have shown that KAP-1 mutants with the ability to bind KRAB but unable to bind HP1 leads to random distribution of KRAZ1 and strong transcriptional activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 SUMO-protein ligase TRIM28; KRAB-A-interacting protein; KRAB-interacting p; KRIP-1; RING-type E3 ubiquitin transferase TIF1-beta; tif1 beta; TIF1-beta; Transcription intermediary factor 1-beta; transcriptional intermediary factor 1, beta; tripartite motif; tripartite motif protein 28; Tripartite motif-containing protein 28
Gene Aliases: AA408787; KAP-1; Kap1; KRIP-1; Krip1; MommeD9; Tif1b; Tif1beta; Trim28
UniProt ID: (Mouse) Q62318
Entrez Gene ID: (Mouse) 21849
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support