Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: big potassium channel alpha subunit; BK alpha; BK channel; BK channel alpha subunit; BKCA alpha; BKCA alpha subunit; bSlo; calcium activated potassium channel protein; calcium-activated potassium channel alpha subunit; Calcium-activated potassium channel subunit alpha-1; Calcium-activated potassium channel, subfamily M subunit alpha-1; hypothetical protein; K(VCA)alpha; KCa1.1; large conductance calcium-activated potassium channel alpha subunit; Maxi K channel; maxi-K channel alpha subunit; maxi-K channel HSLO; MaxiK; MaxiK alpha subunit; MaxiK calcium-activated potassium channel; mSlo1; potassium channel, calcium activated large conductance subfamily M alpha, member 1; potassium large conductance calcium-activated channel, subfamily M, alpha member 1; RbSlo; slo; Slo homolog; Slo-alpha; Slo1; slowpoke; Slowpoke homolog; stretch-activated Kca channel
Gene Aliases: 5730414M22Rik; bA205K10.1; BKCA; BKTM; cSlo; hSlo; I79_002194; KCa1.1; KCNMA; KCNMA1; KCNMA1b; KCNMA1c; MaxiK; mSlo; mSLO1; RBSLO1; SAKCA; SLO; SLO-ALPHA; SLO1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support