Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: MSDYSTGGPP PGPPPPAGGG GGAGGAGGGP PPGPPGAGDR GGGGPGGGGP GGGSAGGPSQ PPGGGGPGIR KDAFADAVQR ARQIAAKIGG DAATTVNNST PDFGFGGQKR QLEDGDQPES KKLASQGDSI SSQLGPIHPP PRTSMTEEYR VPDGMVGLII GRGGEQINKI QQDSGCKVQI
The KHSRP gene encodes a multifunctional RNA-binding protein implicated in a variety of cellular processes, including transcription, alternative pre-mRNA splicing, and mRNA localization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Far upstream element-binding protein 2; FUSE binding protein 2; FUSE-binding protein 2; KH type-splicing regulatory protein; KHE3; KSRP; MAP2 RNA trans-acting protein 1; MAP2 RNA trans-acting protein MARTA1; p75
Gene Aliases: 6330409F21Rik; FBP2; FUBP2; KHSRP; KSRP; Marta1
UniProt ID: (Human) Q92945, (Mouse) Q3U0V1
Entrez Gene ID: (Human) 8570, (Rat) 171137, (Mouse) 16549
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support