Search Thermo Fisher Scientific
PA5-29467 targets KIAA0368 in IF and IHC (P) applications and shows reactivity with Human samples.
The PA5-29467 immunogen is recombinant protein fragment corresponding to a region within amino acids 1727 and 1999 of Human KIAA0368.
Ecm29 appears to be one of three proteins which associate significantly with proteasomes. Ecm29 has been shown to enhance the stability of the proteasome. Ecm29 deletion has been shown to produce 26S proteasomes which dissociated in the absence of ATP. It was thus suggested that Ecm29 tethers the regulatory complex to the proteasome.