Search Thermo Fisher Scientific
Immunogen sequence: MLTAVCGSL GSQHTEAPHA SPPRLDLQPL QTYQGHTSPE AGDYPSPLQP GELQSLPLGP EVDFSQGYEL PGASSRVTCE DLESDSPLAP GPFSKLLQPD MSHHYESWFR PTHPGAEDGS WWDLHPGTSW MDLPHTQGAL TSPGHPGALQ AGLGGYVGDH QLCAPPPHPH AHHLLPAAGG QHLLGPPDGA KALEVAAPES QGLDSSLDGA ARPKGSRRSV PRSSGQTVCR CPNCLE (1-235 aa encoded by B C103951)
SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes.SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes (Scohy et al., 2000).