Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain Serine protease 1; H NRPN; hK8; kallikrein 8; kallikrein 8 (neuropsin/ovasin); Kallikrein-8; kallikrein-related peptidase 8; Kallikrein8; mK8; Neuropsin; NP; Ovasin; PRO322; protease, serine, 19 (neuropsin); Serine protease 19; Serine protease TADG-14; Tumor-associated differentially expressed gene 14 protein; UNQ283
Gene Aliases: BSP1; HNP; KLK8; NP; NRPN; PRSS19; TADG14; UNQ283/PRO322
UniProt ID: (Human) O60259, (Mouse) Q61955
Entrez Gene ID: (Human) 11202, (Dog) 484352, (Rat) 308565, (Mouse) 259277
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support