Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
1 µg/1x10^6 cells | - |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Host/Isotype |
Syrian hamster / IgG |
Class |
Monoclonal |
Type |
Antibody |
Clone |
2F1 |
Immunogen |
Activated NK (A-LAK) cells from B6 (H-2b) mice. |
Conjugate |
FITC |
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Concentration |
0.5 mg/mL |
Purification |
purified |
Storage buffer |
PBS |
Contains |
0.1% sodium azide |
Storage conditions |
4° C, store in dark |
RRID |
AB_2536603 |
KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. KLRG1 is an inhibitory receptor that is expressed on natural killer (NK) cells and certain T cells. NK cells are involved in the lysis of tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: Killer cell lectin-like receptor subfamily G member 1; Mast cell function-associated antigen 2F1; natural killer cell receptor
Gene Aliases: 2F1-Ag; Klrg1; MAFA; MAFA-L
UniProt ID: (Mouse) O88713
Entrez Gene ID: (Mouse) 50928
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support