Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hair keratin; Hair keratin, type I Ha1; hard keratin, type I, 1; K31; keratin 31, type I; keratin, hair, acidic,1; Keratin, type I cuticular Ha1; Keratin-31; KRT hHa1
Gene Aliases: Ha-1; HA1; HHA1; HKA1; KRT31; KRTHA1
UniProt ID: (Human) Q15323
Entrez Gene ID: (Human) 3881
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support