Product References
RNF106 aggravates esophageal squamous cell carcinoma progression through LATS2/YAP axis.
Archives of biochemistry and biophysics
Sun Q,Lu H,Zhang W,Du Y,Liang Q,Zhang Y,Wu J,Zhong M
Sat Jul 01 00:00:00 EDT 2023
RNF106 aggravates esophageal squamous cell carcinoma progression through LATS2/YAP axis.
Archives of biochemistry and biophysics
Sun Q,Lu H,Zhang W,Du Y,Liang Q,Zhang Y,Wu J,Zhong M
Sat Jul 01 00:00:00 EDT 2023
RNF106 aggravates esophageal squamous cell carcinoma progression through LATS2/YAP axis.
Archives of biochemistry and biophysics
Sun Q,Lu H,Zhang W,Du Y,Liang Q,Zhang Y,Wu J,Zhong M
Sat Jul 01 00:00:00 EDT 2023
Pharmacological Activation of YAP/TAZ by Targeting LATS1/2 Enhances Periodontal Tissue Regeneration in a Murine Model.
International journal of molecular sciences
Sato A,Suzuki S,Yuan H,Fahreza RR,Wang X,Nemoto E,Saito M,Yamada S
Wed Jan 04 00:00:00 EST 2023
Pharmacological Activation of YAP/TAZ by Targeting LATS1/2 Enhances Periodontal Tissue Regeneration in a Murine Model.
International journal of molecular sciences
Sato A,Suzuki S,Yuan H,Fahreza RR,Wang X,Nemoto E,Saito M,Yamada S
Wed Jan 04 00:00:00 EST 2023
Arsenic impairs stem cell differentiation via the Hippo signaling pathway.
Toxicology research
Perego MC,McMichael BD,Bain LJ
Sat Apr 01 00:00:00 EDT 2023
O‑GlcNAcylation mediates endometrial cancer progression by regulating the Hippo‑YAP pathway.
International journal of oncology
Zhai L,Yang X,Dong J,Qian L,Gao Y,Lv Y,Chen L,Chen B,Zhou F
Tue Aug 01 00:00:00 EDT 2023
O‑GlcNAcylation mediates endometrial cancer progression by regulating the Hippo‑YAP pathway.
International journal of oncology
Zhai L,Yang X,Dong J,Qian L,Gao Y,Lv Y,Chen L,Chen B,Zhou F
Tue Aug 01 00:00:00 EDT 2023
The m6A methyltransferase Mettl3 deficiency attenuates hepatic stellate cell activation and liver fibrosis.
Molecular therapy : the journal of the American Society of Gene Therapy
Li Y,Kang X,Zhou Z,Pan L,Chen H,Liang X,Chu J,Dong S,Liu C,Yu S,Tu D,Zhang Y,Ge M,Chen W,Xu Y,Zhang Q
Wed Dec 07 00:00:00 EST 2022
The Hippo signaling component LATS2 enhances innate immunity to inhibit HIV-1 infection through PQBP1-cGAS pathway.
Cell death and differentiation
He TS,Dang L,Zhang J,Zhang J,Wang G,Wang E,Xia H,Zhou W,Wu S,Liu X
Sat Jan 01 00:00:00 EST 2022
CircRNA CDR1as promotes cardiomyocyte apoptosis through activating hippo signaling pathway in diabetic cardiomyopathy.
European journal of pharmacology
Shao Y,Li M,Yu Q,Gong M,Wang Y,Yang X,Liu L,Liu D,Tan Z,Zhang Y,Qu Y,Li H,Wang Y,Jiao L,Zhang Y
Thu May 05 00:00:00 EDT 2022
Identifying LATS2 as a prognostic biomarker relevant to immune infiltrates in human esophageal squamous cell carcinoma.
Frontiers in genetics
Zhu M,Liao G,Wang Y,Mo J,Yi D,Zhang Y,Xian L
Tue Sep 20 00:00:00 EDT 2022
SFN Enhanced the Radiosensitivity of Cervical Cancer Cells via Activating LATS2 and Blocking Rad51/MDC1 Recruitment to DNA Damage Site.
Cancers
Wang S,Wang Y,Liu X,Yang Y,Wu S,Liu Y
Fri Apr 08 00:00:00 EDT 2022
SFN Enhanced the Radiosensitivity of Cervical Cancer Cells via Activating LATS2 and Blocking Rad51/MDC1 Recruitment to DNA Damage Site.
Cancers
Wang S,Wang Y,Liu X,Yang Y,Wu S,Liu Y
Fri Apr 08 00:00:00 EDT 2022
Deficiency of large tumor suppressor kinase 1 causes congenital hearing loss associated with cochlear abnormalities in mice.
Biochemical and biophysical research communications
Nishiyama T,Fujioka M,Saegusa C,Oishi N,Harada T,Hosoya M,Saya H,Ogawa K
Fri Jan 01 00:00:00 EST 2021
Tumor-derived exosomal miR-19b-3p facilitates M2 macrophage polarization and exosomal LINC00273 secretion to promote lung adenocarcinoma metastasis via Hippo pathway.
Clinical and translational medicine
Chen J,Zhang K,Zhi Y,Wu Y,Chen B,Bai J,Wang X
Wed Sep 01 00:00:00 EDT 2021
Tumor-derived exosomal miR-19b-3p facilitates M2 macrophage polarization and exosomal LINC00273 secretion to promote lung adenocarcinoma metastasis via Hippo pathway.
Clinical and translational medicine
Chen J,Zhang K,Zhi Y,Wu Y,Chen B,Bai J,Wang X
Wed Sep 01 00:00:00 EDT 2021
CYT387, a potent IKBKE inhibitor, suppresses human glioblastoma progression by activating the Hippo pathway.
Journal of translational medicine
Wang X,Lu J,Li J,Liu Y,Guo G,Huang Q
Mon Sep 20 00:00:00 EDT 2021
CYT387, a potent IKBKE inhibitor, suppresses human glioblastoma progression by activating the Hippo pathway.
Journal of translational medicine
Wang X,Lu J,Li J,Liu Y,Guo G,Huang Q
Mon Sep 20 00:00:00 EDT 2021
CYT387, a potent IKBKE inhibitor, suppresses human glioblastoma progression by activating the Hippo pathway.
Journal of translational medicine
Wang X,Lu J,Li J,Liu Y,Guo G,Huang Q
Mon Sep 20 00:00:00 EDT 2021
CYT387, a potent IKBKE inhibitor, suppresses human glioblastoma progression by activating the Hippo pathway.
Journal of translational medicine
Wang X,Lu J,Li J,Liu Y,Guo G,Huang Q
Mon Sep 20 00:00:00 EDT 2021
Schwannoma development is mediated by Hippo pathway dysregulation and modified by RAS/MAPK signaling.
JCI insight
Chen Z,Li S,Mo J,Hawley E,Wang Y,He Y,Brosseau JP,Shipman T,Clapp DW,Carroll TJ,Le LQ
Thu Oct 15 00:00:00 EDT 2020
Schwannoma development is mediated by Hippo pathway dysregulation and modified by RAS/MAPK signaling.
JCI insight
Chen Z,Li S,Mo J,Hawley E,Wang Y,He Y,Brosseau JP,Shipman T,Clapp DW,Carroll TJ,Le LQ
Thu Oct 15 00:00:00 EDT 2020
The Human Novel Gene LNC-HC Inhibits Hepatocellular Carcinoma Cell Proliferation by Sequestering hsa-miR-183-5p.
Molecular therapy. Nucleic acids
Lan X,Wu N,Wu L,Qu K,Osoro EK,Guan D,Du X,Wang B,Chen S,Miao J,Ren J,Liu L,Li H,Ning Q,Li D,Lu S
Fri Jun 05 00:00:00 EDT 2020
TRIB3 confers radiotherapy resistance in esophageal squamous cell carcinoma by stabilizing TAZ.
Oncogene
Zhou S,Liu S,Lin C,Li Y,Ye L,Wu X,Jian Y,Dai Y,Ouyang Y,Zhao L,Liu M,Song L,Xi M
Wed Apr 01 00:00:00 EDT 2020
Cholesterol Stabilizes TAZ in Hepatocytes to Promote Experimental Non-alcoholic Steatohepatitis.
Cell metabolism
Wang X,Cai B,Yang X,Sonubi OO,Zheng Z,Ramakrishnan R,Shi H,Valenti L,Pajvani UB,Sandhu J,Infante RE,Radhakrishnan A,Covey DF,Guan KL,Buck J,Levin LR,Tontonoz P,Schwabe RF,Tabas I
Tue May 05 00:00:00 EDT 2020
Hippo pathway and tumoral FOXP3 expression correlate with tumor growth in squamous cell carcinoma of the lung.
Pathology, research and practice
Won KY,Kim HK,Kim GY,Song MJ,Lim SJ
Wed Jul 01 00:00:00 EDT 2020
MicroRNA‑93 promotes angiogenesis and attenuates remodeling via inactivation of the Hippo/Yap pathway by targeting Lats2 after myocardial infarctionω.
Molecular medicine reports
Ma C,Peng P,Zhou Y,Liu T,Wang L,Lu C
Wed Jul 01 00:00:00 EDT 2020
CLDN6 promotes tumor progression through the YAP1-snail1 axis in gastric cancer.
Cell death & disease
Yu S,Zhang Y,Li Q,Zhang Z,Zhao G,Xu J
Wed Dec 11 00:00:00 EST 2019
Essential Oil from Pinus Koraiensis Pinecones Inhibits Gastric Cancer Cells via the HIPPO/YAP Signaling Pathway.
Molecules (Basel, Switzerland)
Zhang Y,Xin C,Qiu J,Wang Z
Fri Oct 25 00:00:00 EDT 2019
The CalcR-PKA-Yap1 Axis Is Critical for Maintaining Quiescence in Muscle Stem Cells.
Cell reports
Zhang L,Noguchi YT,Nakayama H,Kaji T,Tsujikawa K,Ikemoto-Uezumi M,Uezumi A,Okada Y,Doi T,Watanabe S,Braun T,Fujio Y,Fukada SI
Tue Nov 19 00:00:00 EST 2019
Myricetin Suppresses the Propagation of Hepatocellular Carcinoma via Down-Regulating Expression of YAP.
Cells
Li M,Chen J,Yu X,Xu S,Li D,Zheng Q,Yin Y
Wed Apr 17 00:00:00 EDT 2019
miR-25 Promotes Cell Proliferation, Migration, and Invasion of Non-Small-Cell Lung Cancer by Targeting the LATS2/YAP Signaling Pathway.
Oxidative medicine and cellular longevity
Wu T,Hu H,Zhang T,Jiang L,Li X,Liu S,Zheng C,Yan G,Chen W,Ning Y,Li Y,Lu Z
Mon Jan 06 00:00:00 EST 2020
miR-25 Promotes Cell Proliferation, Migration, and Invasion of Non-Small-Cell Lung Cancer by Targeting the LATS2/YAP Signaling Pathway.
Oxidative medicine and cellular longevity
Wu T,Hu H,Zhang T,Jiang L,Li X,Liu S,Zheng C,Yan G,Chen W,Ning Y,Li Y,Lu Z
Mon Jan 06 00:00:00 EST 2020
Large tumor suppressor 2, LATS2, activates JNK in a kinase-independent mechanism through ASK1.
Journal of molecular cell biology
Rusnak L,Tang C,Qi Q,Mo X,Fu H
Sat Dec 01 00:00:00 EST 2018
Large tumor suppressor 2, LATS2, activates JNK in a kinase-independent mechanism through ASK1.
Journal of molecular cell biology
Rusnak L,Tang C,Qi Q,Mo X,Fu H
Sat Dec 01 00:00:00 EST 2018
A comprehensive evaluation of Hippo pathway silencing in sarcomas.
Oncotarget
Merritt NM,Fullenkamp CA,Hall SL,Qian Q,Desai C,Thomason J,Lambertz AM,Dupuy AJ,Darbro B,Tanas MR
Fri Aug 03 00:00:00 EDT 2018
A comprehensive evaluation of Hippo pathway silencing in sarcomas.
Oncotarget
Merritt NM,Fullenkamp CA,Hall SL,Qian Q,Desai C,Thomason J,Lambertz AM,Dupuy AJ,Darbro B,Tanas MR
Fri Aug 03 00:00:00 EDT 2018
A comprehensive evaluation of Hippo pathway silencing in sarcomas.
Oncotarget
Merritt NM,Fullenkamp CA,Hall SL,Qian Q,Desai C,Thomason J,Lambertz AM,Dupuy AJ,Darbro B,Tanas MR
Fri Aug 03 00:00:00 EDT 2018
A comprehensive evaluation of Hippo pathway silencing in sarcomas.
Oncotarget
Merritt NM,Fullenkamp CA,Hall SL,Qian Q,Desai C,Thomason J,Lambertz AM,Dupuy AJ,Darbro B,Tanas MR
Fri Aug 03 00:00:00 EDT 2018
YAP triggers the Wnt/β-catenin signalling pathway and promotes enterocyte self-renewal, regeneration and tumorigenesis after DSS-induced injury.
Cell death & disease
Deng F,Peng L,Li Z,Tan G,Liang E,Chen S,Zhao X,Zhi F
Fri Feb 02 00:00:00 EST 2018
YAP triggers the Wnt/β-catenin signalling pathway and promotes enterocyte self-renewal, regeneration and tumorigenesis after DSS-induced injury.
Cell death & disease
Deng F,Peng L,Li Z,Tan G,Liang E,Chen S,Zhao X,Zhi F
Fri Feb 02 00:00:00 EST 2018
MicroRNA-31 activates the RAS pathway and functions as an oncogenic MicroRNA in human colorectal cancer by repressing RAS p21 GTPase activating protein 1 (RASA1).
The Journal of biological chemistry
Sun D,Yu F,Ma Y,Zhao R,Chen X,Zhu J,Zhang CY,Chen J,Zhang J
Fri Mar 29 00:00:00 EDT 2013
MicroRNA-31 activates the RAS pathway and functions as an oncogenic MicroRNA in human colorectal cancer by repressing RAS p21 GTPase activating protein 1 (RASA1).
The Journal of biological chemistry
Sun D,Yu F,Ma Y,Zhao R,Chen X,Zhu J,Zhang CY,Chen J,Zhang J
Fri Mar 29 00:00:00 EDT 2013