Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
LHPP is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Phospholysine phosphohistidine inorganic pyrophosphate phosphatase
Gene Aliases: 2310007H09Rik; Lhpp
UniProt ID: (Mouse) Q9D7I5, (Rat) Q5I0D5
Entrez Gene ID: (Pig) 100156052, (Mouse) 76429, (Rat) 361663
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support