Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
81% identical in mouse, 80% identical in rat. <30% identity with LRRK1.
10 µg/mL of MA5-45642 was sufficient for detection of LRRK2/Dardarin in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects >200kDa.
This antibody was formerly sold as clone S231B-34.
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. Mutations in LRRK2 have been associated with Parkinson's disease. LRRK2 is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: augmented in rheumatoid arthritis 17; BC 300-268; Dardarin; DKFZp434H2111; FLJ45829; leucine-rich repeat kinase 2; Leucine-rich repeat serine/threonine-protein kinase 2; LRR kinase 2; LRRK 2; LRRK-2
Gene Aliases: AURA17; DARDARIN; LRRK2; PARK8; RIPK7; ROCO2
UniProt ID: (Human) Q5S007
Entrez Gene ID: (Human) 120892, (Rat) 300160
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support