Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 3
PA1-331B detects recombinant human liver X receptor (LXR).
PA1-331B has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~64 kDa and ~80 kDa protein representing recombinant human LXR alpha and beta.
The PA1-331B immunogen is a synthetic peptide corresponding to residues C L(432) R L Q D K K L P P L L S E(445) of human LXR alpha. This sequence also corresponds to amino acids 442-455 of the human LXR beta. PA1-331B immunizing peptide (Cat. # PEP-115) is available for use in neutralization and control experiments.
NR1H3 (oxysterols receptor LXR-alpha) is a nuclear receptor that exhibits a ligand-dependent transcriptional activiation activity. Its interaction with retinoic acid receptor (RXR) shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4-type response elements characterized by direct repeats of two similar hexanucleotide half-sites spaced by four nucleotides. NR1H3 plays an important role in the regulation of cholesterol homeostasis, regulating cholesterol uptake through MYLIP-dependent ubiqutination of LDLR, VLDLR, and LRP8. It also interplays functionally with RORA for the regulation of genes involves in liver metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: liver X nuclear receptor alpha variant 1; liver X nuclear receptor beta; Liver X Receptor; Liver X receptor alpha; Liver X receptor beta; LX receptor beta; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 2; nuclear receptor subfamily 1, group H, member 3; Oxysterols receptor LXR-alpha; Oxysterols receptor LXR-beta; steroid hormone-nuclear receptor NER; Ubiquitously-expressed nuclear receptor
Gene Aliases: LXR-a; LXR-b; LXRA; LXRB; NER; NER-I; NR1H2; NR1H3; RIP15; RLD-1; UNR
UniProt ID: (Human) P55055, (Human) Q13133
Entrez Gene ID: (Human) 7376, (Human) 10062
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support