Product References
Neuregulin-1-human epidermal receptor-2 signaling is a central regulator of pulmonary epithelial permeability and acute lung injury.
The Journal of biological chemistry
Finigan JH,Faress JA,Wilkinson E,Mishra RS,Nethery DE,Wyler D,Shatat M,Ware LB,Matthay MA,Mason R,Silver RF,Kern JA
MA5-12893 was used in western blot to study the role of neuregulin-1 and HER2 in regulating pulmonary epithelial permeability and acute lung injury
Fri Mar 25 00:00:00 EDT 2011
Phenotypic characterization of transgenic mice overexpressing neuregulin-1.
PloS one
Kato T,Kasai A,Mizuno M,Fengyi L,Shintani N,Maeda S,Yokoyama M,Ozaki M,Nawa H
MA5-12893 was used in western blot to study the cognitive and behavioural phenotype of transgenic mice overexpressing neuregulin-1
Thu Dec 09 00:00:00 EST 2010
Neuregulin-1 signaling in brain endothelial cells.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Lok J,Sardi SP,Guo S,Besancon E,Ha DM,Rosell A,Kim WJ,Corfas G,Lo EH
MA5-12893 was used in western blot to study neuregulin-1 signaling in brain microvascular endothelial cells
Thu Jan 01 00:00:00 EST 2009
Ventricular ErbB2/ErbB4 activation and downstream signaling in pacing-induced heart failure.
Journal of molecular and cellular cardiology
Doggen K,Ray L,Mathieu M,Mc Entee K,Lemmens K,De Keulenaer GW
MA5-12893 was used in western blot to investigate the involvement of ventricular ErbB2 and ErbB4 receptors in the development of pacing-induced heart failure
Thu Jan 01 00:00:00 EST 2009
Peripheral neuropathy in the twitcher mouse: accumulation of extracellular matrix in the endoneurium and aberrant expression of ion channels.
Acta neuropathologica
Kagitani-Shimono K,Mohri I,Yagi T,Taniike M,Suzuki K
MA5-12893 was used in immunohistochemistry to investigate the extracellular and protein abnormalities in peripheral neuropathy
Thu May 01 00:00:00 EDT 2008
Constitutive neuregulin-1/ErbB signaling contributes to human vestibular schwannoma proliferation.
Glia
Hansen MR,Roehm PC,Chatterjee P,Green SH
MA5-12893 was used in immunoprecipitation and western blot to study the role of constitutive neuregulin-1/ErbB signaling in human vestibular schwannoma proliferation
Sat Apr 15 00:00:00 EDT 2006
Localization of neuregulin-1alpha (heregulin-alpha) and one of its receptors, ErbB-4 tyrosine kinase, in developing and adult human brain.
Brain research bulletin
Bernstein HG,Lendeckel U,Bertram I,Bukowska A,Kanakis D,Dobrowolny H,Stauch R,Krell D,Mawrin C,Budinger E,Keilhoff G,Bogerts B
MA5-12893 was used in immunohistochemistry to study the localization of neuregulin-1alpha and ErbB-4 tyrosine kinase in developing and adult human brain
Mon May 15 00:00:00 EDT 2006
Role of neuregulin-1/ErbB2 signaling in endothelium-cardiomyocyte cross-talk.
The Journal of biological chemistry
Lemmens K,Segers VF,Demolder M,De Keulenaer GW
MA5-12893 was used in immunoprecipitation to study the role of neuregulin-1/ErbB2 signaling in endothelium-cardiomyocyte cross-talk
Fri Jul 14 00:00:00 EDT 2006
NRG1 gene rearrangements in clinical breast cancer: identification of an adjacent novel amplicon associated with poor prognosis.
Oncogene
Prentice LM,Shadeo A,Lestou VS,Miller MA,deLeeuw RJ,Makretsov N,Turbin D,Brown LA,Macpherson N,Yorida E,Cheang MC,Bentley J,Chia S,Nielsen TO,Gilks CB,Lam W,Huntsman DG
MA5-12893 was used in immunohistochemistry to investigate the association of NRG1 gene rearrangements with poor prognosis in breast cancer treatment
Thu Nov 10 00:00:00 EST 2005
Neuregulin-1 enhances survival of human astrocytic glioma cells.
Glia
Ritch PS,Carroll SL,Sontheimer H
MA5-12893 was used in western blot to study the mechanism by which neuregulin-1 promotes the survival of human astrocytoma cells
Mon Aug 15 00:00:00 EDT 2005
Paclitaxel and carboplatin as first-line chemotherapy combined with gefitinib (IRESSA) in patients with advanced breast cancer: a phase I/II study conducted by the Hellenic Cooperative Oncology Group.
Breast cancer research and treatment
Fountzilas G,Pectasides D,Kalogera-Fountzila A,Skarlos D,Kalofonos HP,Papadimitriou C,Bafaloukos D,Lambropoulos S,Papadopoulos S,Kourea H,Markopoulos C,Linardou H,Mavroudis D,Briasoulis E,Pavlidis N,Razis E,Kosmidis P,Gogas H
MA5-12893 was used in immunohistochemistry to investigate the efficacy of the combination of paclitaxel, carboplatin and gefitinib against advanced breast cancer
Fri Jul 01 00:00:00 EDT 2005
Expression of heregulin in human coronary atherosclerotic lesions.
Journal of vascular research
Panutsopulos D,Arvanitis DL,Tsatsanis C,Papalambros E,Sigala F,Spandidos DA
MA5-12893 was used in immunohistochemistry to study the expression of heregulin in human coronary atherosclerotic lesions
Mon Dec 05 00:00:00 EST 2005
Back signaling by the Nrg-1 intracellular domain.
The Journal of cell biology
Bao J,Wolpowitz D,Role LW,Talmage DA
MA5-12893 was used in immunocytochemistry to study the bidirectional signaling of neuregulin-1 in neurones and the role of the intracellular domain in retrograde signaling
Mon Jun 23 00:00:00 EDT 2003
Reciprocal signaling between spiral ganglion neurons and Schwann cells involves neuregulin and neurotrophins.
Hearing research
Hansen MR,Vijapurkar U,Koland JG,Green SH
MA5-12893 was used in immunocytochemistry to study the role of neuregulin and neurotrophins in regulating spiral ganglion neuron-Schwann cell reciprocal signaling in the auditory nerve
Thu Nov 01 00:00:00 EST 2001