Product References
Elevated glucose increases genomic instability by inhibiting nucleotide excision repair.
Life science alliance
Ciminera AK,Shuck SC,Termini J
Published figure using XPA monoclonal antibody (Product # MA5-13835) in Western Blot
Fri Oct 01 00:00:00 EDT 2021
Progerin sequestration of PCNA promotes replication fork collapse and mislocalization of XPA in laminopathy-related progeroid syndromes.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Hilton BA,Liu J,Cartwright BM,Liu Y,Breitman M,Wang Y,Jones R,Tang H,Rusinol A,Musich PR,Zou Y
Published figure using XPA monoclonal antibody (Product # MA5-13835) in Western Blot
Fri Sep 01 00:00:00 EDT 2017
ATR pathway inhibition is synthetically lethal in cancer cells with ERCC1 deficiency.
Cancer research
Mohni KN,Kavanaugh GM,Cortez D
MA5-13835 was used in western blot to study the the synthetic lethality of ATR inhibitors in cells deficient for ERCC1 and the implications for cancer therapy
Thu May 15 00:00:00 EDT 2014
A rapid assay for measuring nucleotide excision repair by oligonucleotide retrieval.
Scientific reports
Shen JC,Fox EJ,Ahn EH,Loeb LA
MA5-13835 was used in blocking or activating experiment to develop an assay for the sensitive and quantitative measuremt of nucleotide excision repair activity in a variety of human cell types
Thu May 08 00:00:00 EDT 2014
Physiological modulation of endogenous BRCA1 p220 abundance suppresses DNA damage during the cell cycle.
Genes & development
Dimitrov SD,Lu D,Naetar N,Hu Y,Pathania S,Kanellopoulou C,Livingston DM
MA5-13835 was used in immunocytochemistry and western blot to study the importance of correct physiological regulation of BRCA1 p220 levels in maintaining genome stability through the cell cycle
Tue Oct 15 00:00:00 EDT 2013
Lack of CAK complex accumulation at DNA damage sites in XP-B and XP-B/CS fibroblasts reveals differential regulation of CAK anchoring to core TFIIH by XPB and XPD helicases during nucleotide excision repair.
DNA repair
Zhu Q,Wani G,Sharma N,Wani A
MA5-13835 was used in immunocytochemistry to study the role of the XPB and XPD helicases in regulating the anchorage of the CAK complex to TFIIH during nucleotide excision repair
Sat Dec 01 00:00:00 EST 2012
Sodium arsenite and hyperthermia modulate cisplatin-DNA damage responses and enhance platinum accumulation in murine metastatic ovarian cancer xenograft after hyperthermic intraperitoneal chemotherapy (HIPEC).
Journal of ovarian research
Muenyi CS,States VA,Masters JH,Fan TW,Helm CW,States JC
MA5-13835 was used in immunocytochemistry to report that sodium arsenite and hyperthermia modulate cisplatin-DNA damage responses and enhance platinum accumulation in murine metastatic ovarian cancer xenograft after hyperthermic intraperitoneal chemothera
Wed Jun 22 00:00:00 EDT 2011
Dissociation of CAK from core TFIIH reveals a functional link between XP-G/CS and the TFIIH disassembly state.
PloS one
Arab HH,Wani G,Ray A,Shah ZI,Zhu Q,Wani AA
MA5-13835 was used in immunocytochemistry to study the functional association between XP-G/CS mutations and the disassembly state of TFIIH
Tue Jun 08 00:00:00 EDT 2010
Chromatin restoration following nucleotide excision repair involves the incorporation of ubiquitinated H2A at damaged genomic sites.
DNA repair
Zhu Q,Wani G,Arab HH,El-Mahdy MA,Ray A,Wani AA
MA5-13835 was used in immunocytochemistry to study the role of ubiquitinated H2A in chromatin restoration after nucleotide excision repair
Sun Feb 01 00:00:00 EST 2009
Recognition of cisplatin-DNA interstrand cross-links by replication protein A.
Biochemistry
Patrick SM,Tillison K,Horn JM
MA5-13835 was used in western blot to study the ability of replication protein to recognize and bind to cisplatin-DNA interstrand cross-links
Tue Sep 23 00:00:00 EDT 2008
Molecular characterization of spontaneous mesenchymal stem cell transformation.
PloS one
Rubio D,Garcia S,Paz MF,De la Cueva T,Lopez-Fernandez LA,Lloyd AC,Garcia-Castro J,Bernad A
MA5-13835 was used in western blot to investigate the molecular mechanisms of spontaneous mesenchymal stem cell transformation
Wed Jan 02 00:00:00 EST 2008
DNA polymerase epsilon associates with the elongating form of RNA polymerase II and nascent transcripts.
The FEBS journal
Rytkönen AK,Hillukkala T,Vaara M,Sokka M,Jokela M,Sormunen R,Nasheuer HP,Nethanel T,Kaufmann G,Pospiech H,Syväoja JE
MA5-13835 was used in immunoprecipitation to study the interaction of DNA polymerase epsilon with the elongating form of RNA polymerase II
Fri Dec 01 00:00:00 EST 2006
Expression of xeroderma pigmentosum A protein predicts improved outcome in metastatic ovarian carcinoma.
Cancer
Stevens EV,Raffeld M,Espina V,Kristensen GB,Trope' CG,Kohn EC,Davidson B
MA5-13835 was used in immunohistochemistry to evaluate the prognostic value of xeroderma pigmentosum A protein expression in metastatic ovarian carcinoma
Wed Jun 01 00:00:00 EDT 2005
Functional implications of antiestrogen induction of quinone reductase: inhibition of estrogen-induced deoxyribonucleic acid damage.
Molecular endocrinology (Baltimore, Md.)
Bianco NR,Perry G,Smith MA,Templeton DJ,Montano MM
MA5-13835 was used in immunocytochemistry to study the role of quinone reductase induction by anti-estrogens in protecting against oxidative DNA damage by estrogen metabolites
Tue Jul 01 00:00:00 EDT 2003