    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Isotype Controls ›
          • Mouse IgG1 Isotype Controls
          Invitrogen
          Mouse IgG1 Recombinant Isotype Control (HyHEL-10)
          View all Mouse IgG1 Isotype Controls
          Questions & Answers
          Cite Mouse IgG1 Recombinant Isotype Control (HyHEL-10)
          Additional Information:
          {{banner.description}}
          • Testing Data (1)
          Mouse IgG1 Isotype Control in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          Mouse IgG1 Isotype Control in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Mouse IgG1 Isotype Control (MA5-55126) in SDS-PAGE
          SDS-PAGE analysis of Mouse IgG1. Samples were run under reducing (R) and non-reducing (NR) conditions using Mouse IgG1 (Product # MA5-55126) monoclonal antibody. The purity of the protein is greater than 95%. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mouse IgG1 Isotype Control in SDS-PAGE (SDS-PAGE)
          Product Details
          MA5-55126
          Applications
          Tested Dilution
          Flow Cytometry (Flow)
          Assay-dependent
          Product Specifications
          Host/Isotype
          Mouse / IgG1
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Isotype Control
          Clone
          HyHEL-10
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS
          Contains
          no preservative
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          RRID
          AB_3667372
          Target Information
          Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
          View product citations for antibody MA5-55126 on CiteAb
          Bioinformatics
          Protein Aliases: Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline