Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMAdCAM-1; MAdCAM-1; Mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1
Gene Aliases: MACAM1; MADCAM1
UniProt ID: (Human) Q13477
Entrez Gene ID: (Human) 8174
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support