Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17121 targets MESP2 in indirect ELISA and WB applications and shows reactivity with Human samples.
The MA5-17121 immunogen is purified recombinant fragment of human MESP2 (amino acids: 37-94 ) expressed in E. Coli.
MA5-17121 detects MESP2 which has a predicted molecular weight of approximately 41.8kDa.
This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHc6; Class C basic helix-loop-helix protein 6; mesoderm posterior 2 homolog; mesoderm posterior basic helix-loop-helix transcription factor 2; Mesoderm posterior protein 2
Gene Aliases: BHLHC6; MESP2; SCDO2
UniProt ID: (Human) Q0VG99
Entrez Gene ID: (Human) 145873
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support