Search Thermo Fisher Scientific
MA1-27176 detects HLA DR beta from human samples.
MA1-27176 has been successfully used in immunohistochemistry (paraffin), and Western blot procedures.
The MA1-27176 immunogen is P815 transfectant expressing a human/mouse hybrid molecule: human HLADR4b1 domain in a mouse IE background.
The protein encoded by this gene is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The encoded membrane protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in this gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms have been found for this gene. Other variants may also exist, but the full-length natures of their transcripts has yet to be defined.