Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Transcriptional repressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mesoderm induction early response 1, family member 2; Mesoderm induction early response protein 2; Mi-er2
Gene Aliases: KIAA1193; Mi-er2; MIER2
UniProt ID: (Human) Q8N344
Entrez Gene ID: (Human) 54531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support