Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Panda, Dog, Horse, Rabbit (100%) Hamster, Elephant, Bovine, Bat, Pig (94%).
MAP3K9 (MLK1) is a serine/threonine protein kinase that functions in the MAP kinase pathway. Plays an important role in the cascades of cellular responses evoked by changes in the environment. Once activated, acts as an upstream activator of the MKK/JNK signal transduction cascade through the phosphorylation of MAP2K4/MKK4 and MAP2K7/MKK7 which in turn activate the JNKs. The MKK/JNK signaling pathway regulates stress response via activator protein-1 (JUN) and GATA4 transcription factors. Plays also a role in mitochondrial death signaling pathway, including the release cytochrome c, leading to apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitogen activated protein kinase kinase kinase 9; Mitogen-activated protein kinase kinase kinase 9; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity)
Gene Aliases: E130314H24Rik; MAP3K9; MEKK9; MLK1; PRKE1
UniProt ID: (Human) P80192, (Mouse) Q3U1V8
Entrez Gene ID: (Dog) 490765, (Horse) 100054289, (Human) 4293, (Rabbit) 100341731, (Mouse) 338372
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support