Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: ALLEPGAGVL SIYYLGKKFL GDLQPDGRIM WQETGQTFNS PSAWATHCKK LVNPAKKSGC GWASVKYKGQ KLDKYKATWL RLHQLHTPAT AADESPAS
Probable protease. Acts as a sensor of N(6)-methyladenosine methylation on DNA (m6A): recognizes and binds m6A DNA, leading to its degradation (PubMed:30982744). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ14981; MPN domain-containing protein
Gene Aliases: MPND
UniProt ID: (Human) Q8N594
Entrez Gene ID: (Human) 84954
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support