Search Thermo Fisher Scientific
Immunogen sequence: IHSCESISL GLSTFMIDLN QQVAKAVNNA TAQSLVLIDE FGKGTNTVDG LALLAAVLRH WLARGPTCPH IFVATNFLSL VQLQLLPQGP LVQYLTMETC EDGNDLVFFY QVCEGVAKAS HASHTAAQAG LPDKLVARGK EVSDLIRSGK PIKPVKDLLK KNQMENCQTL VDKFMKLDLE DPNLDLNVFM SQEVLPAATS IL (635-835 aa encoded by BC002498)
MSH5 is a member of the mutS family of proteins that are involved in DNA mismatch repair or meiotic recombination processes. This protein is similar to a Saccharomyces cerevisiae protein that participates in meiotic segregation fidelity and crossing-over. This protein forms heterooligomers with another member of this family, mutS homolog 4. This gene encodes a member of the mutS family of proteins that are involved in DNA mismatch repair or meiotic recombination processes. This protein is similar to a Saccharomyces cerevisiae protein that participates in meiotic segregation fidelity and crossing-over. This protein forms heterooligomers with another member of this family, mutS homolog 4. Alternative splicing results in four transcript variants encoding three different isoforms.