Search Thermo Fisher Scientific
PA1-4175 detects MUC12 in human samples.
PA1-4175 has been successfully used in Western blot procedures.
The PA1-4175 immunogen is a synthetic peptide corresponding to residues S(10) S I S G E P T S L Y S Q A E(24) of human MUC12.
Epitheial mucins are secreted glycoproteins that play roles in cell protection, adhesion modulation and signaling. MUC12 is a novel mucin gene and may be involved in epithelial cell growth regulation. MUC12 is down-regulated in colorectal cancers.