Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAGGPGAGS AAPVSSTSSL PLAALNMRVR RRLSLFLNVR TQVAADWTAL AEEMDFEYLE IRQLETQADP TGRLLDAWQG RPGASVGRLL ELLTKLGRDD VLLELGPSIE EDCQKYILKQ QQEEAEKPLQ VAAVDSSVPR TAELAGITTL DDPLGHMPER FDAFICYCPS DIQFVQEMIR QLEQTNYRLK LCVSDRDVLP GTCVWSIASE LIEKRCRRMV VVVSDDYLQS KECDFQTKFA LSLSPGAHQK RLIPIKYKAM KKEFPSILRS ITVCDYTNPC TKSWFWTRLA KALSLP
Myeloid differentiation Marker 88 (MyD88) is an adaptor molecule for the Toll/IL-1R and is involved in the inflammatory response induced by IL-1, IL-18 and LPS. MyD88 recruits IRAK to IL-1 receptor complex in response to IL-1. This pathway further leads to activation of NF-kB. MyD88 has a wide tissue distribution.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mutant myeloid differentiation primary response 88; myeloid differentiation primary response gene (88); myeloid differentiation primary response gene 88; Myeloid differentiation primary response protein MyD88
Gene Aliases: MYD88; MYD88D
UniProt ID: (Human) Q99836, (Rat) Q6Y1S1
Entrez Gene ID: (Human) 4615, (Rat) 301059
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support