Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A1 catalytic; A2 catalytic; MLC1/MLC3; MLC1F/MLC3F; Myosin light chain 1/3, skeletal muscle isoform; Myosin light chain A1/A2; Myosin light chain alkali 1/2; myosin, light chain 1, alkali; skeletal, fast; myosin, light polypeptide 1, alkali; skeletal, fast
Gene Aliases: MLC1F; MLC3F; MYL1
UniProt ID: (Human) P05976
Entrez Gene ID: (Human) 4632
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support