Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in both slow-twitch skeletal muscle and in nonmuscle tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MLC1sa; myl6b; myosin alkali light chain 1 slow a; myosin light chain 1 slow a; Myosin light chain 1 slow-twitch muscle A isoform; myosin light chain 1, slow-twitch muscle A isoform; Myosin light chain 6B; myosin, light chain 6B, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle; sm; smooth muscle and non-muscle myosin alkali light chain 6B; Smooth muscle and nonmuscle myosin light chain alkali 6B
Gene Aliases: 5730437E04Rik; BC037527; MLC1SA; MYL6B
UniProt ID: (Human) P14649, (Mouse) Q8CI43
Entrez Gene ID: (Human) 140465, (Mouse) 216459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support