Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 20 kDa myosin light chain; LC20; MGC3505; MLC-2C; myosin light chain, regulatory C; myosin regulatory light chain; myosin regulatory light chain 1; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light polypeptide 9; Myosin RLC; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory
Gene Aliases: AI327049; LC20; MLC-2C; MLC2; MLC20; MRLC1; MYL9; Mylc2c; MYRL2; RLC-C
UniProt ID: (Human) P24844, (Rat) Q64122, (Mouse) Q9CQ19
Entrez Gene ID: (Human) 10398, (Rat) 296313, (Mouse) 98932
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support