Search Thermo Fisher Scientific
The MA1-7393 antibody reacts with Marburg virus in viral and infected tissue samples.
MA1-7393 has been successfully used in ELISA and Western blot applications.
The MA1-7393 antibody was raised against Marburg virus 30 kDa antigen.
Marburg virus (MBG) is a representative of Filoviridae family of RNA containing viruses. MBG is an exceptionally dangerous pathogen, which induces a severe contagious and highly lethal (53-88%) febrile disease with hemorrhagic syndrome. The name "thread viruses" is based on their morphology. MBG made its appearance in 1967 in the form of frightening nosocomial outbreak, initially among polio vaccine production workers in Germany in contact with Ugandan green monkeys and their kidney tissues. The patho-and immuno-genesis of MBG fever have so far been little studied, no specific agents or treatment methods have been developed.