Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The monoclonal antibody 25F9 recognizes a protein on mature macrophages both on the cell surface and in intracellular vesicular structures. Expression is thought to correlate with endocytic macrophages and is useful in many disease models. Expression is not found on immature macrophages or monocytes or any other hematopoietic cell.
Applications Reported: This eBio25F9 (25F9) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), immunohistology staining of frozen and paraffin embedded tissue sections.
Applications Tested: This eBio25F9 (25F9) antibody has been tested by flow cytometric analysis of human macrophages. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ED2-like antigen; Macrophages; Mature Macrophages
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support