Search Thermo Fisher Scientific
The MA1-7426 antibody specifically detects Measles virus HA protein in viral and infected tissue samples.
MA1-7426 has been successfully used in ELISA and immunofluorescence applications.
The MA1-7426 antibody was raised against a viral preparation.