Product References
Xp11.2 translocation renal cell carcinoma with NONO-TFE3 gene fusion: morphology, prognosis, and potential pitfall in detecting TFE3 gene rearrangement.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Xia QY,Wang Z,Chen N,Gan HL,Teng XD,Shi SS,Wang X,Wei X,Ye SB,Li R,Ma HH,Lu ZF,Zhou XJ,Rao Q
MA134864 was used in immunohistochemistry - paraffin section to characterize eight cases of renal cell carcinoma cases with NONO-TFE3
Wed Mar 01 00:00:00 EST 2017
Primary sclerosing epithelioid fibrosarcoma of kidney with variant histomorphologic features: report of 2 cases and review of the literature.
Diagnostic pathology
Ertoy Baydar D,Kosemehmetoglu K,Aydin O,Bridge JA,Buyukeren B,Aki FT
MA1-34864 was used in immunohistochemistry to present two cases of primary sclerosing epithelioid fibrosarcoma of the kidney
Fri Oct 09 00:00:00 EDT 2015
Loss of BRM expression is a frequently observed event in poorly differentiated clear cell renal cell carcinoma.
Histopathology
Xia QY,Rao Q,Cheng L,Shen Q,Shi SS,Li L,Liu B,Zhang J,Wang YF,Shi QL,Wang JD,Ma HH,Lu ZF,Yu B,Zhang RS,Zhou XJ
MA1-34864 was used in immunohistochemistry to study the incidence of BRM expression loss in patients with poorly differentiated clear cell renal cell carcinoma
Thu May 01 00:00:00 EDT 2014
Perivascular epithelioid cell tumor (PEComa) with TFE3 gene rearrangement: clinicopathological, immunohistochemical, and molecular features.
Virchows Archiv : an international journal of pathology
Shen Q,Rao Q,Xia QY,Yu B,Shi QL,Zhang RS,Zhou XJ
MA1-34864 was used in immunohistochemistry - paraffin section to describe three cases of perivascular epithelioid cell tumors with transcription factor E3 gene abnormalities
Sat Nov 01 00:00:00 EDT 2014
Cathepsin K expression in a wide spectrum of perivascular epithelioid cell neoplasms (PEComas): a clinicopathological study emphasizing extrarenal PEComas.
Histopathology
Rao Q,Cheng L,Xia QY,Liu B,Li L,Shi QL,Shi SS,Yu B,Zhang RS,Ma HH,Lu ZF,Tu P,Zhou XJ
MA1-34864 was used in immunohistochemistry to study the expression and diagnostic value of cathepsin K expression in a wide range of extra-renal perivascular epithelioid cell neoplasms
Fri Mar 01 00:00:00 EST 2013
Renal cell carcinomas with t(6;11)(p21;q12) presenting with tubulocystic renal cell carcinoma-like features.
International journal of clinical and experimental pathology
Rao Q,Zhang XM,Tu P,Xia QY,Shen Q,Zhou XJ,Shi QL
MA1-34864 was used in immunohistochemistry to report on a case of renal cell carcinoma with t(6;11)(p21;q12
Fri Feb 07 00:00:00 EST 2014
Renal cell carcinomas with t(6;11)(p21;q12): A clinicopathologic study emphasizing unusual morphology, novel alpha-TFEB gene fusion point, immunobiomarkers, and ultrastructural features, as well as detection of the gene fusion by fluorescence in situ hybridization.
The American journal of surgical pathology
Rao Q,Liu B,Cheng L,Zhu Y,Shi QL,Wu B,Jiang SJ,Wang Y,Wang X,Yu B,Zhang RS,Ma HH,Lu ZF,Tu P,Wang JD,Zhou XJ
MA1-34864 was used in immunohistochemistry to perform a detailed clinicopathological study of renal cell carcinomas with t(6;11)(p21;q12)
Sat Sep 01 00:00:00 EDT 2012
Expression of E-cadherin in angiomyolipoma.
Human pathology
Wang Z,Gong Q,Fan Q
MA1-34864 was used in immunohistochemistry to perform a systematic study of E-cadherin expression in angiomyolipoma
Sat Dec 01 00:00:00 EST 2012
Sporadic haemangioblastoma of the kidney with rhabdoid features and focal CD10 expression: report of a case and literature review.
Diagnostic pathology
Yin WH,Li J,Chan JK
MA1-34864 was used in immunohistochemistry to report on a case of sporadic haemangioblastoma of the kidney with rhabdoid features and focal CD10 expression
Thu Apr 12 00:00:00 EDT 2012
Microscopic uterine lymphangioleiomyomatosis perivascular epithelioid cell neoplasm: a case report with the earliest manifestation of this enigmatic neoplasm.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists
Clay MR,Gibson P,Lowell J,Cooper K
MA1-34864 was used in immunohistochemistry to report a clinical case of microscopic lymphangioleiomyomatosis perivascular epithelioid cell neoplasm in uterus
Sat Jan 01 00:00:00 EST 2011
Diagnostic challenges of metastatic spindle cell melanoma on fine-needle aspiration specimens.
Cancer
Piao Y,Guo M,Gong Y
MA1-34864 was used in immunohistochemistry to review the diagnostic difficulty of metastatic spindle cell melanoma
Fri Apr 25 00:00:00 EDT 2008
Melanocytes expressing MC1R polymorphisms associated with red hair color have altered MSH-ligand activated pigmentary responses in coculture with keratinocytes.
Journal of cellular physiology
Roberts DW,Newton RA,Leonard JH,Sturm RA
MA1-34864 was used in western blot to study the behavior and functional abilities of melanocytes expressing MC1R red hair color variants
Thu May 01 00:00:00 EDT 2008
Hyperpigmentation during interferon-alpha therapy for chronic hepatitis C virus infection.
The British journal of dermatology
Willems M,Munte K,Vrolijk JM,Den Hollander JC,Böhm M,Kemmeren MH,De Man RA,Brouwer JT
MA1-34864 was used in immunohistochemistry to study melanin deposition during interferon-alpha therapy for HCV infection
Fri Aug 01 00:00:00 EDT 2003
Acral lentiginous melanoma: an immunohistochemical study of 20 cases.
International journal of dermatology
Kim YC,Lee MG,Choe SW,Lee MC,Chung HG,Cho SH
MA1-34864 was used in immunohistochemistry to identify the diagnostic markers for acral lentiginous melanoma
Sat Feb 01 00:00:00 EST 2003
Comparison of five antibodies as markers in the diagnosis of melanoma in cytologic preparations.
American journal of clinical pathology
Sheffield MV,Yee H,Dorvault CC,Weilbaecher KN,Eltoum IA,Siegal GP,Fisher DE,Chhieng DC
MA1-34864 was used in immunohistochemistry to study 5 antibodies as diagnostic markers of melanoma incytologic preparation
Sun Dec 01 00:00:00 EST 2002