Search Thermo Fisher Scientific
Working Concentration: 0.3125 µg/mL. Sensitivity: 10 ng/mL Meloxicam Sodium salt produces 78% inhibition in a competitive ELISA employing Meloxicam polyclonal antibody.
Meloxicam is an oxicam derivative and a non-steroidal anti-inflammatory drug (NSAID) with anti-inflammatory, antipyretic and analgesic activities. Unlike traditional nonselective NSAIDs, meloxicam preferentially inhibits the activity of cyclo-oxygenase II (COX-II), resulting in a decreased conversion of arachidonic acid into prostaglandin precursors. The resulting decrease in prostaglandin synthesis is responsible for the therapeutic effects of meloxicam. [National Center for Biotechnology Information. PubChem Compound Database; CID=54677470].
Protein Aliases: C14H13N3O4S2; Meloxicamum; Mobic; Mobicox; Movalis