Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This antibody is specific for the matrix protein of human metapneumovirus. It does not cross-react with Respiratory Syncytial Virus, Influenza A, Influenza B, Adenovirus or Parainfluenza viruses 1, 2 & 3.
Purity is >90%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h-MPV; hMPV; hMPV matrix; MPV matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support