Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution |
---|---|
Western Blot (WB) |
Assay-dependent |
ELISA (ELISA) |
5 µg/mL as capture antibody |
Product Specifications | |
---|---|
Species Reactivity |
Rat |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Secondary Antibody |
Clone |
MARE-1 |
Immunogen |
IR162, IR1016, IR2 and IR410 rat IgE myeloma proteins |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS |
Contains |
0.1% sodium azide |
Storage conditions |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
RRID |
AB_2538293 |
Target |
IgE |
Antibody Form |
Whole Antibody |
Mouse anti Rat IgE antibody, clone MARE-1 recognizes rat epsilon heavy chain of immunoglobulin IgE and does not cross-react with other classes of rat immunoglobulin.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support