Search Thermo Fisher Scientific
MA5-12339 targets c-myb/v-myb in IHC (P) and WB applications and shows reactivity with Human samples.
The MA5-12339 immunogen is recombinant v-myb protein.
Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5'-YAAC[GT]G-3'. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.