Product References
Primary mouse myoblast metabotropic purinoceptor profiles and calcium signalling differ with their muscle origin and are altered in mdx dystrophinopathy.
Scientific reports
Róg J,Oksiejuk A,Górecki DC,Zabłocki K
MA5-12902 was used in Immunocytochemistry-immunofluorescence to identify significant differences in the pattern of expression and activity of P2Y receptors and the levels of the calcium signalling toolkit" proteins between mdx and wild-type myoblasts isolated from different muscles."
Thu Jun 08 00:00:00 EDT 2023
Integrative molecular roadmap for direct conversion of fibroblasts into myocytes and myogenic progenitor cells.
Science advances
Kim I,Ghosh A,Bundschuh N,Hinte L,Petrosyan E,von Meyenn F,Bar-Nur O
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Immunocytochemistry
Fri Apr 08 00:00:00 EDT 2022
MSTN Regulatory Network in Mongolian Horse Muscle Satellite Cells Revealed with miRNA Interference Technologies.
Genes
Budsuren U,Ulaangerel T,Shen Y,Liu G,Davshilt T,Yi M,Bold D,Zhang X,Bai D,Dorjgotov D,Davaakhuu G,Jambal T,Li B,Du M,Dugarjav M,Bou G
MA5-12902 was used in Immunohistochemistry to study the regulatory mechanism of the transforming growth factor-β member, myostatin (MSTN), in equine muscle cells.
Tue Oct 11 00:00:00 EDT 2022
MSTN Regulatory Network in Mongolian Horse Muscle Satellite Cells Revealed with miRNA Interference Technologies.
Genes
Budsuren U,Ulaangerel T,Shen Y,Liu G,Davshilt T,Yi M,Bold D,Zhang X,Bai D,Dorjgotov D,Davaakhuu G,Jambal T,Li B,Du M,Dugarjav M,Bou G
MA5-12902 was used in Immunohistochemistry to study the regulatory mechanism of the transforming growth factor-β member, myostatin (MSTN), in equine muscle cells.
Tue Oct 11 00:00:00 EDT 2022
Simple and effective serum-free medium for sustained expansion of bovine satellite cells for cell cultured meat.
Communications biology
Stout AJ,Mirliani AB,Rittenberg ML,Shub M,White EC,Yuen JSK,Kaplan DL
MA5-12902 was used in Immunocytochemistry-immunoflourescence to offer a simple, affordable, and effective Beefy-9 and Beefy-9+ serum-free media for improving cultured meat research.
Thu Jun 02 00:00:00 EDT 2022
Upregulation of miR181a/miR212 Improves Myogenic Commitment in Murine Fusion-Negative Rhabdomyosarcoma.
Frontiers in physiology
Pozzo E,Giarratana N,Sassi G,Elmastas M,Killian T,Wang CC,Marini V,Ronzoni F,Yustein J,Uyttebroeck A,Sampaolesi M
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Immunocytochemistry
Tue Aug 24 00:00:00 EDT 2021
Upregulation of miR181a/miR212 Improves Myogenic Commitment in Murine Fusion-Negative Rhabdomyosarcoma.
Frontiers in physiology
Pozzo E,Giarratana N,Sassi G,Elmastas M,Killian T,Wang CC,Marini V,Ronzoni F,Yustein J,Uyttebroeck A,Sampaolesi M
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Immunocytochemistry
Tue Aug 24 00:00:00 EDT 2021
FOXP1 Interacts with MyoD to Repress its Transcription and Myoblast Conversion.
Journal of cellular signaling
Wright WE,Li C,Zheng CX,Tucker HO
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Western Blot
Thu Feb 11 00:00:00 EST 2021
Myo/Nog cells expressing muscle proteins are present in preretinal membranes from patients with proliferative vitreoretinopathy.
Experimental eye research
Gerhart J,Morrison N,Gugerty L,Telander D,Bravo-Nuevo A,George-Weinstein M
MA5-12902 was used in Immunohistochemistry to investigate the function of Myo/Nog cells in three ocular trauma patients with proliferative vitreoretinopathy.
Sat Aug 01 00:00:00 EDT 2020
Myo/Nog cells are nonprofessional phagocytes.
PloS one
Gerhart J,Gugerty L,Lecker P,Abdalla F,Martin M,Gerhart O,Gerhart C,Johal K,Bernstein J,Spikes J,Mathers K,Bravo-Nuevo A,George-Weinstein M
MA5-12902 was used in Immunocytochemistry-immunoflourescence to present evidence suggesting that Myo/Nog cells phagocytose tattoo ink in tissue sections of human skin and engulf cell corpses in cultures of anterior human lens tissue and magnetic beads injected into the anterior chamber of mice in vivo.
Wed Sep 30 00:00:00 EDT 2020
Myo/Nog cells are nonprofessional phagocytes.
PloS one
Gerhart J,Gugerty L,Lecker P,Abdalla F,Martin M,Gerhart O,Gerhart C,Johal K,Bernstein J,Spikes J,Mathers K,Bravo-Nuevo A,George-Weinstein M
MA5-12902 was used in Immunocytochemistry-immunoflourescence to present evidence suggesting that Myo/Nog cells phagocytose tattoo ink in tissue sections of human skin and engulf cell corpses in cultures of anterior human lens tissue and magnetic beads injected into the anterior chamber of mice in vivo.
Wed Sep 30 00:00:00 EDT 2020
Brain-specific angiogenesis inhibitor 1 is expressed in the Myo/Nog cell lineage.
PloS one
Gerhart J,Bowers J,Gugerty L,Gerhart C,Martin M,Abdalla F,Bravo-Nuevo A,Sullivan JT,Rimkunas R,Albertus A,Casta L,Getts L,Getts R,George-Weinstein M
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Immunohistochemistry
Tue Sep 08 00:00:00 EDT 2020
Brain-specific angiogenesis inhibitor 1 is expressed in the Myo/Nog cell lineage.
PloS one
Gerhart J,Bowers J,Gugerty L,Gerhart C,Martin M,Abdalla F,Bravo-Nuevo A,Sullivan JT,Rimkunas R,Albertus A,Casta L,Getts L,Getts R,George-Weinstein M
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Immunohistochemistry
Tue Sep 08 00:00:00 EDT 2020
Rhabdomyosarcoma and Wilms tumors contain a subpopulation of noggin producing, myogenic cells immunoreactive for lens beaded filament proteins.
PloS one
Gerhart J,Behling K,Paessler M,Milton L,Bramblett G,Garcia D,Pitts M,Hurtt R,Crawford M,Lackman R,Nguyen D,Infanti J,FitzGerald P,George-Weinstein M
MA5-12902 was used in Immunohistochemistry-immunofluorescence to screen rhabdomyosarcoma and Wilms tumours for a subpopulation of noggin producing, myogenic cells immunoreactive for lens beaded filament proteins.
Thu Dec 19 00:00:00 EST 2019
Rhabdomyosarcoma and Wilms tumors contain a subpopulation of noggin producing, myogenic cells immunoreactive for lens beaded filament proteins.
PloS one
Gerhart J,Behling K,Paessler M,Milton L,Bramblett G,Garcia D,Pitts M,Hurtt R,Crawford M,Lackman R,Nguyen D,Infanti J,FitzGerald P,George-Weinstein M
MA5-12902 was used in Immunohistochemistry-immunofluorescence to screen rhabdomyosarcoma and Wilms tumours for a subpopulation of noggin producing, myogenic cells immunoreactive for lens beaded filament proteins.
Thu Dec 19 00:00:00 EST 2019
Isolation, cultivation and immunostaining of single myofibers: An improved approach to study the behavior of satellite cells.
Journal of biological methods
Lim CL,Ling KH,Cheah PS
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Immunocytochemistry
Sat Nov 04 00:00:00 EDT 2023
PAX3-FOXO1 transgenic zebrafish models identify HES3 as a mediator of rhabdomyosarcoma tumorigenesis.
eLife
Kendall GC,Watson S,Xu L,LaVigne CA,Murchison W,Rakheja D,Skapek SX,Tirode F,Delattre O,Amatruda JF
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Western Blot
Tue Jun 05 00:00:00 EDT 2018
Inflammation-associated miR-155 activates differentiation of muscular satellite cells.
PloS one
Onodera Y,Teramura T,Takehara T,Itokazu M,Mori T,Fukuda K
MA5-12902 was used in Western Blotting to study the expression of microRNA-155, in muscular satellite cells of aged muscles.
Fri Mar 22 00:00:00 EDT 2019
The RNA Surveillance Factor UPF1 Represses Myogenesis via Its E3 Ubiquitin Ligase Activity.
Molecular cell
Feng Q,Jagannathan S,Bradley RK
MA5-12902 was used in Western Blotting to study the role of UPF1 in regulated protein decay.
Thu Jul 20 00:00:00 EDT 2017
Genetic disruption of Smad7 impairs skeletal muscle growth and regeneration.
The Journal of physiology
Cohen TV,Kollias HD,Liu N,Ward CW,Wagner KR
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Immunofluorescence
Mon Jun 01 00:00:00 EDT 2015
Genetic disruption of Smad7 impairs skeletal muscle growth and regeneration.
The Journal of physiology
Cohen TV,Kollias HD,Liu N,Ward CW,Wagner KR
Published figure using MYOD monoclonal antibody (Product # MA5-12902) in Immunofluorescence
Mon Jun 01 00:00:00 EDT 2015
Molecular characterization of an EWSR1-POU5F1 fusion associated with a t(6;22) in an undifferentiated soft tissue sarcoma.
Cancer genetics
Deng FM,Galvan K,de la Roza G,Zhang S,Souid AK,Stein CK
MA5-12902 was used in immunohistochemistry to characterize a clinical case of undifferentiated soft tissue sarcoma with chromosomal rearrangement and resultant EWSR1-POU5F1 fusion
Mon Aug 01 00:00:00 EDT 2011
CD90-positive cells, an additional cell population, produce laminin alpha2 upon transplantation to dy(3k)/dy(3k) mice.
Experimental cell research
Fukada S,Yamamoto Y,Segawa M,Sakamoto K,Nakajima M,Sato M,Morikawa D,Uezumi A,Miyagoe-Suzuki Y,Takeda S,Tsujikawa K,Yamamoto H
MA5-12902 was used in immunocytochemistry to study the role of CD90-positive cells in producing laminin alpha2 upon transplantation to dy(3k)/dy(3k) mice
Tue Jan 01 00:00:00 EST 2008
Molecular signature of quiescent satellite cells in adult skeletal muscle.
Stem cells (Dayton, Ohio)
Fukada S,Uezumi A,Ikemoto M,Masuda S,Segawa M,Tanimura N,Yamamoto H,Miyagoe-Suzuki Y,Takeda S
MA5-12902 was used in immunocytochemistry to investigate gene expression in quiescent satellite cells of adult skeletal muscle
Mon Oct 01 00:00:00 EDT 2007
ADP-ribosylation of integrin alpha7 modulates the binding of integrin alpha7beta1 to laminin.
The Biochemical journal
Zhao Z,Gruszczynska-Biegala J,Zolkiewska A
MA5-12902 was used in western blot to study the effect of ADP-ribosylation on the interaction between integrin alpha7beta1 and laminin
Sat Jan 01 00:00:00 EST 2005
Roles of peroxisome proliferator-activated receptors delta and gamma in myoblast transdifferentiation.
Experimental cell research
Holst D,Luquet S,Kristiansen K,Grimaldi PA
MA5-12902 was used in western blot to study the roles of PPARdelta and gamma in myoblast transdifferentiation
Fri Aug 01 00:00:00 EDT 2003