Search Thermo Fisher Scientific
MA1-24940 detects Myosin light chain from human, bovine, chicken, porcine, and rabbit samples.
MA1-24940 has been successfully used in Western blot and immunofluorescence procedures.
The MA1-24940 immunogen is chicken lens membranes.
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.