Invitrogen
MA1-10774 detects N-acetylprocainamide.
MA1-10774 has been successfully used in ELISA applications.
The MA1-10774 immunogen is N-Acetylprocainamide, conjugated to KLH.
N-Acetylprocainamide (NAPA or acecainide) is the N-acetylated metabolite of procainamide. It is a Class III antiarrhythmic agent, whereas procainamide is a Class Ia antiarrhythmic drug. It is only partially as active as procainamide and may cause cardiac toxicity and kidney failure. [National Center for Biotechnology Information. PubChem Compound Database; CID=4342].