Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: YQLTLWGPEG NILDYANKQL AGLVANYYTP RWRLFLEALV DSVAQGIPFQ QHQFDKNVFQ LEQAFVLSKQ RYPSQPRGDT VDLAKKIFLK YYPGWVAGS
This gene encodes an enzyme that degrades heparan sulfate by hydrolysis of terminal N-acetyl-D-glucosamine residues in N-acetyl-alpha-D-glucosaminides. Defects in this gene are the cause of mucopolysaccharidosis type IIIB (MPS-IIIB), also known as Sanfilippo syndrome B. This disease is characterized by the lysosomal accumulation and urinary excretion of heparan sulfate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-N-acetylglucosaminidase; alpha-N-acetylglucosaminidase, lysosomal; N-acetyl-alpha-glucosaminidase; N-acetyl-glucosaminidase; N-acetylglucosaminidase, alpha; NAG; testicular tissue protein Li 18
Gene Aliases: CMT2V; MPS-IIIB; MPS3B; NAG; NAGLU; UFHSD; UFHSD1
UniProt ID: (Human) P54802
Entrez Gene ID: (Human) 4669, (Mouse) 27419
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support