Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MLRTAGRDGL CRLSTYLEEL EAVELKKFKL YLGTATELGE GKIPWGSMEK AGPLEMAQLL ITHFGPEEAW RLALSTFERI NRKDLWERGQ REDLVRDTPP GGPSSLGNQS
NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP12, also known as Monarch-1, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. NALP12 is thought to act as an attenuating factor of inflammation by suppressing inflammatory responses such as NF-kappa-B activation by TLR-signaling molecules MyD88, IRAK-1, TRAF6 and RIPK1 in activated monocytes. Recent evidence suggests that mutations in NALP12 result in hereditary periodic fever syndromes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: monarch 1; Monarch-1; NACHT, LRR and PYD domains-containing protein 12; NLR family, pyrin domain containing 12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 12; PYRIN-containing APAF1-like protein 7; Regulated by nitric oxide
Gene Aliases: CLR19.3; FCAS2; NALP12; NLRP12; PAN6; PYPAF7; RNO; RNO2
UniProt ID: (Human) P59046
Entrez Gene ID: (Human) 91662
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support