Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:5,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Reptile |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Recombinant Naja kaouthia Cobra venom factor protein (733-984aa) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
5 mg/mL |
Purification |
Protein G |
Storage buffer |
10mM PBS, pH 7.4, with 50% glycerol |
Contains |
0.03% ProClin 300 |
Storage conditions |
-20°C or -80°C if preferred |
RRID |
AB_2902334 |
This antibody has been tested in direct-ELISA
Complement-activating protein in cobra venom. It is a structural and functional analog of complement component C3b, the activated form of C3. It binds factor B (CFB), which is subsequently cleaved by factor D (CFD) to form the bimolecular complex CVF/Bb. CVF/Bb is a C3/C5 convertase that cleaves both complement components C3 and C5. Structurally, it resembles the C3b degradation product C3c, which is not able to form a C3/C5 convertase. Unlike C3b/Bb, CVF/Bb is a stable complex and completely resistant to the actions of complement regulatory factors H (CFH) and I (CFI). Therefore, CVF continuously activates complement resulting in the depletion of complement activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CVF
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support