Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects endogenous protein at a molecular weight of 62 kDa.
Purity is >95% by SDS-PAGE.
NEURL is involved in the determination of cell fate in the neurogenic region of the embryo and plays a role in the determination of cell fate in the central nervous system. NEURL may act as a tumor suppressor whose inactivation could be associated with malignant progression of astrocytic tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase NEURL1; h neu; h neuralized 1; h-neu; h-neuralized 1; neuralized homolog; neuralized homolog 1A; neuralized-like homolog; Neuralized-like protein 1A; RING finger protein 67; RING-type E3 ubiquitin transferase NEURL1
Gene Aliases: bA416N2.1; neu; neu-1; NEUR1; NEURL; NEURL1; NEURL1A; RNF67
UniProt ID: (Human) O76050
Entrez Gene ID: (Human) 9148, (Rat) 309459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support