Search Thermo Fisher Scientific
Glycerol (1:1) may be added for additional stability.
Reconstitute in 100 µL of sterile water.
The peptide is homologous in rat.
VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.